Learn More
Medchemexpress LLC Petrolatum (white) | 8009-03-8 | MFCD00147839 | ≥99.0% | 500g
SDP
Catalog No. 5000269885
Encompass_Preferred
Petrolatum (white) is an ointment vehicle for many agents due to its low skin irritation Petrolatum (white) is a safe effective wound care ointment for ambulatory surgery[1][2][3]
Catalog No. 50-002-69885 Supplier Medchemexpress LLC Supplier No. HYY0683A500G
Petrolatum (white) is an ointment vehicle for many agents due to its low skin irritation Petrolatum (white) is a safe effective wound care ointment for ambulatory surgery[1][2][3]