Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, 20° to 40°C, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003580 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Aluminum Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-580 4 L Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-580 Supplier Avantor Supplier No. 927203
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 8030-30-6
Synonym benzol, cyclohexatriene, benzole, pyrobenzole, benzine, phenyl hydride, pyrobenzol, benzen, phene, mineral naphtha

Specifications

Quantity 4 L
Flash Point −40.00°C
Packaging Aluminum Bottle
Specific Gravity 1L = 0.63kg
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Petroleum Ether, 20° to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.