Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, 35° to 60°C, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003620 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-620 4 L Aluminum Bottle
02-003-591 1 L Aluminum Bottle
02-003-621 4 L Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-003-620 Supplier Avantor Supplier No. 926803
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 8032-32-4
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Quantity 4 L
Flash Point −18.00°C
Packaging Aluminum Bottle
Specific Gravity 1L = 0.68kg
Grade ACS
Chemical Name or Material Petroleum Ether, 35° to 60°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.