Learn More
Sigma Aldrich Fine Chemicals Biosciences Petroleum ether ACS reagen
Catalog No. 502897804
Encompass
Petroleum ether is a mixture of low boiling aliphatic hydrocarbons (mainly hexane and pentane) that is generally used as a laboratory solvent.
Catalog No. 50-289-7804 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1845192.5L
Petroleum ether is a mixture of low boiling aliphatic hydrocarbons (mainly hexane and pentane) that is generally used as a laboratory solvent.