Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RSOP02004C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOP02004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOP02004C Supplier Ricca Chemical Company Supplier No. RSOP02004C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Ligroin)

Chemical Identifiers

CAS 8032-32-4

Specifications

Acidity Pass
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.001%
Boiling Point 35°C to 60°C
Grade ACS
Chemical Name or Material Petroleium Ether
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.