Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, ACS Reagent (Ligroin), Reagents

Catalog No. 01337999
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Glass Bottle
Steel Pail
Tin Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-999 1 L Glass Bottle
01-337-997 20 L Steel Pail
01-337-996 4 L Glass Bottle
01-337-994 10 L Tin Can
01-337-993 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-999 Supplier Reagents Holdings Llc Supplier No. C11990001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Petroleum Ether, ACS Reagent (Ligroin), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Petroleum Ether, ACS Reagent (Ligroin)
CAS 8032-32-4
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.645
Molecular Formula C6-16H12-34
Synonym Naphtha, Petroleum benzine, Petroleum spirit, Ligroin, Ligroine
Boiling Point 35°C
Grade ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.