Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether Analytical Grade, Lab Chem

Catalog No. LC181204 Shop All LabChem Products
Change view
Click to view available options
Boiling Point:
35 °C to 60 °C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Boiling Point
LC181204 35 °C to 60 °C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC181204 Supplier LabChem, Inc. Supplier No. LC181204
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Petroleum Ether Analytical Grade Solvent and Reagent, Safety: Flammable

Chemical Identifiers

CAS 8032-32-4
Molecular Weight (g/mol) 90

Specifications

Quantity 4 L
Packaging Amber Glass
Boiling Point 35 °C to 60 °C
Percent Purity >90%
Grade Analytical
Chemical Name or Material Petroleum Ether Analytical Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.