Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether BP, 65 to 100°C, MilliporeSigma™

Catalog No. 50947966 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-966 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-947-966 Supplier MilliporeSigma Supplier No. PX04273
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 8032-32-4

Specifications

Quantity 4 L
Flash Point -7°C
Packaging Glass Bottle, Pre-Packed
Residue after Evaporation 0.001% max
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.