Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum ether, extra pure, boiling range 100-140°C

Catalog No. ac233020100
Change view
Click to view available options
Quantity:
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233020100 10 L Plastic Drum
AC233020025 2.5 L Glass Bottle
AC233020250 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC233020100 Supplier Thermo Scientific Chemicals Supplier No. 233020100
Only null left
Add to Cart

CAS: 64742-49-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Petroleum ether
Melting Point -30.0°C
CAS 64742-49-0, 111-65-9, 142-82-5, 108-87-2, 110-82-7, 73513-42-5
Density 0.7400g/mL
Quantity 10 L
Flash Point 2°C
Infrared Spectrum Authentic
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.4030 to 1.4080
Residue after Evaporation 0.005% max.
Specific Gravity 0.74
Viscosity 0.76 mm2/s (25°C)
MDL Number MFCD00081849
Merck Index 14, 5489
Boiling Point 100.0°C to 140.0°C
Solubility Information Solubility in water: insoluble.
IUPAC Name Petroleum ether
Grade Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.