Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Petroleum Ether, Liquid
SCI_ED

Catalog No. S25461B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 lb.
Packaging:
ACS Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25461B 1 lb. ACS Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25461B

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 8032-32-4
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Quantity 1 lb.
Packaging ACS Grade
Physical Form Liquid
Grade ACS
Chemical Name or Material Petroleum Ether
Safety handling Solvent
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.