Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, Purified (Ligroin), Reagents

Catalog No. 01337268
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-268 500 mL
01-337-269 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-268 Supplier Reagents Holdings Llc Supplier No. C2199000500C
Add to Cart
Add to Cart

Petroleum Ether, Purified (Ligroin), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 8032-32-4
Molecular Formula C6-16H12-34
Synonym Naphtha, Petroleum benzine, Petroleum spirit, Ligroin, Ligroine

Specifications

Color Colorless
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.645
Boiling Point 35°C to 60°C
Grade Pure
Chemical Name or Material Petroleum Ether, Purified (Ligroin)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.