Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0% (C5 and C6 Isomers), Solstice

Catalog No. 6001173 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
Packaging:
Metal Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-011-73 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-011-73 Supplier Solstice Supplier No. PP31719
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8032-32-4
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

CAS Max % 100.0000%
Density 0.64 g/cm3
Quantity 19 L
Flash Point −18°C
Packaging Metal Can
Water 0.02% max.
UN Number UN1268
Boiling Point 35°C to 60°C
Percent Purity >99%
Grade ACS
Assay by GC total C5 and C6 isomers
Chemical Name or Material Petroleum Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0% (C5 & C6 Isomers)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Petroleum distillates, n.o.s.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.