Learn More
Sigma Aldrich Fine Chemicals Biosciences Petroleum ether puriss. p.1L
Catalog No. 502897867
Encompass
Petroleum ether is a low boiling point fraction having boiling point in the range of 40-60C. Extracts of leaves of Litchi chinensis Gaertn. (Sapindaceae) have been prepared in petroleum ether and were evaluated for antiinflammatory analgesic and antipyretic activities.
Catalog No. 50-289-7867 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 322991L
Petroleum ether is a low boiling point fraction having boiling point in the range of 40-60C. Extracts of leaves of Litchi chinensis Gaertn. (Sapindaceae) have been prepared in petroleum ether and were evaluated for antiinflammatory analgesic and antipyretic activities.