Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroleum Ether, Spectrophotometric Grade, Solstice

Catalog No. 60048546 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-546 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-546 Supplier Solstice Supplier No. 2617341L

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8032-32-4
MDL Number MFCD00081849
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Density 0.64g/mL at 25°C
Quantity 1 L
Explosion Limit 0.08
Flash Point −49°C (−56.2°F)
Ignition Point 475°F
Packaging Glass Bottle
Vapor Pressure 25.8psi (55°C), 7.99psi (20°C)
UN Number UN1268
Boiling Point 40°C to 60°C
Grade Spectrophotometry
Chemical Name or Material Petroleum Ether
Show More Show Less
Hazard Statement P201-P210-P280-P308 + P313-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H340-H350
DOTInformation class 3 - PG 1 - Petroleum distillates, n.o.s., HI: no
EINECSNumber 232-453-7
RTECSNumber OI6180000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.