Learn More
Grainger Petroleum Jelly 13 oz Tub
Catalog No. 502748816
Encompass_Preferred
The Grainger PETROLEUM JELLY 13 oz Tub is a petroleum-based jelly that can be used as a lubricant, protectant, or moisturizer. It is made in the USA and contains 13 ounces of USP grade petroleum jelly. The product has a melting point of 100-140°F and is triple purified to be odorless and colorless. It can be used on skin, lips, and hair and helps prevent chapping and irritation. The tub keeps the jelly free of contaminants. Overall, this US-made petroleum jelly provides versatile protection and lubrication.
Catalog No. 50-274-8816 Supplier Grainger Supplier No. 14U287
The Grainger PETROLEUM JELLY 13 oz Tub is a petroleum-based jelly that can be used as a lubricant, protectant, or moisturizer. It is made in the USA and contains 13 ounces of USP grade petroleum jelly. The product has a melting point of 100-140°F and is triple purified to be odorless and colorless. It can be used on skin, lips, and hair and helps prevent chapping and irritation. The tub keeps the jelly free of contaminants. Overall, this US-made petroleum jelly provides versatile protection and lubrication.