Learn More
Cayman Chemical Petromurin C, Purity: ≥95%, CAS: 194608-29-2, 2.5 mg
SDP
Catalog No. 502934471
Encompass_Preferred
A fungal metabolite; cytotoxic to NS-1 mouse myeloma cells (IC50 = 33 μg/ml); has activity against T. foetus in vitro (IC50 = 100 μg/ml)
Catalog No. 50-293-4471 Supplier Cayman Chemical Supplier No. 250892.5 MG
A fungal metabolite; cytotoxic to NS-1 mouse myeloma cells (IC50 = 33 μg/ml); has activity against T. foetus in vitro (IC50 = 100 μg/ml)