Learn More
Medchemexpress LLC HY-113362 10mg Medchemexpress, Petroselinic acid CAS:593-39-5 Purity:>98%
SDP
Catalog No. 501969246
Encompass_Preferred
Medchemexpress, HY-113362 10mg Petroselinic acid CAS:593-39-5 Petroselinic acid, a positional isomer of oleic acid, is isolated from the vegetable oil of Coriandrum sativum fruits. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9246 Supplier Medchemexpress LLC Supplier No. HY11336210MG
Medchemexpress, HY-113362 10mg Petroselinic acid CAS:593-39-5 Petroselinic acid, a positional isomer of oleic acid, is isolated from the vegetable oil of Coriandrum sativum fruits. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.