Learn More
Selleck Chemical LLC Petroselinic acid S3342-5mg
SDP
Supplier:  Selleck Chemical LLC S33425MG
Encompass_Preferred
Petroselinic acid (PSA) is a positional isomer of oleic acid isolated from the vegetable oil of Coriandrum sativum fruits Petroselinic acid can be used for the fermentation of new sophorolipids
Catalog No. 50-310-9993