Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroselinic Acid, ≥99%, MP Biomedicals™

Catalog No. ICN15611491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15611491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15611491 Supplier MP Biomedicals, Inc Supplier No. 0215611491

missing translation for 'validMainframeMsg'

Specifications

CAS 593-39-5
Color Colorless to White
Assay Percent Range ≥99%
Molecular Formula C18H34O2
Quantity 1 g
MDL Number MFCD00046561
Synonym petroselinic acid, cis-6-octadecenoic acid, z-octadec-6-enoic acid, petroselic acid, 6-octadecylenic acid, 6-octadecenoic acid, z, petroselinsaeure, 6z-octadecenoic acid, petroseleninsaeure, 6z-6-octadecenoic acid
InChI Key CNVZJPUDSLNTQU-SEYXRHQNSA-N
SMILES CCCCCCCCCCC\C=C/CCCCC(O)=O
IUPAC Name (6Z)-octadec-6-enoic acid
Molecular Weight (g/mol) 282.47
PubChem CID 5281125
ChEBI CHEBI:28194
Formula Weight 282.5
Physical Form Powder to Solid
Chemical Name or Material Petroselinic Acid
Show More Show Less
EINECSNumber 209-789-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.