Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petroselinic Acid 98.0+%, TCI America™
SDP

Catalog No. O00091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O00091G Supplier TCI America Supplier No. O00091G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 593-39-5
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00046561
InChI Key CNVZJPUDSLNTQU-SEYXRHQNSA-N
Synonym petroselinic acid, cis-6-octadecenoic acid, z-octadec-6-enoic acid, petroselic acid, 6-octadecylenic acid, 6-octadecenoic acid, z, petroselinsaeure, 6z-octadecenoic acid, petroseleninsaeure, 6z-6-octadecenoic acid
PubChem CID 5281125
ChEBI CHEBI:28194
IUPAC Name (6Z)-octadec-6-enoic acid
SMILES CCCCCCCCCCC\C=C/CCCCC(O)=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 282.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Petroselinic Acid
EINECSNumber (2)-0609
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.