Learn More
Cayman Chemical PetunIdInchlorIde 10mg
SDP
Catalog No. 502984357
Encompass_Preferred
An O-methylated anthocyanidin that imparts blue-red pigments to flowers, fruits, and red wine; suppresses the activity of focal adhesion kinase and the proliferation of HCT116 colon cancer cells
Catalog No. 50-298-4357 Supplier Cayman Chemical Supplier No. 1975510 MG
An O-methylated anthocyanidin that imparts blue-red pigments to flowers, fruits, and red wine; suppresses the activity of focal adhesion kinase and the proliferation of HCT116 colon cancer cells