Learn More
Chemie Tek MLN4924 (PEVONEDISTAT), 905579-51-3, MFCD17215201, 5mg
SDP
Supplier:  Chemie Tek CTM49245MG
Encompass_Preferred
Molecular Formula: C21H25N5O4S ∙ H2O, 99.9-100% pure by achiral and chiral HPLCs, MW: 461.5; MLN4924 is a small molecule inhibitor of the NEDD8-Activating Enzyme (NAE), a key component of the protein homeostasis pathway. It is the first small molecule inhibitor specifically targeting this class of enzyme to be studied clinically, and is currently being examined in Phase I clinical trials.
Catalog No. NC0792192