Learn More
Medchemexpress LLC Pevonedistat hydrochloride | 1160295-21-5 | 98.6% | 479.98 g·mol⁻¹ | C21H26ClN5O4S | 100 MG
SDP
Catalog No. 5000243420
Encompass_Preferred
Pevonedistat hydrochloride is a potent and selective NEDD8-activating enzyme (NAE) inhibitor used in research to study ubiquitin-like protein modification, protein turnover, and cell-cycle regulation. It is supplied as a white to off-white solid with high analytical purity and is intended for laboratory research and analytical applications. CAS 1160295-21-5.
  • Potent, selective inhibitor of NAE for mechanistic studies.
  • High reported purity suitable for analytical use.
  • White to off-white solid appearance for solid-phase handling.
  • Stable when stored sealed and protected from moisture.
  • In solvent: store at -80°C (up to 1 year) or -20°C (up to 6 months).
Catalog No. 50-002-43420 Supplier Medchemexpress LLC Supplier No. HY10484100MG
Pevonedistat hydrochloride is a potent and selective NEDD8-activating enzyme (NAE) inhibitor used in research to study ubiquitin-like protein modification, protein turnover, and cell-cycle regulation. It is supplied as a white to off-white solid with high analytical purity and is intended for laboratory research and analytical applications. CAS 1160295-21-5.
  • Potent, selective inhibitor of NAE for mechanistic studies.
  • High reported purity suitable for analytical use.
  • White to off-white solid appearance for solid-phase handling.
  • Stable when stored sealed and protected from moisture.
  • In solvent: store at -80°C (up to 1 year) or -20°C (up to 6 months).