Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pevonedistat hydrochloride | 1160295-21-5 | MFCD17215670 | 99.2% | 480.0 g/mol | C21H26ClN5O4S | 50 MG
SDP

Catalog No. 5000242009
Encompass_Preferred

Pevonedistat hydrochloride is the hydrochloride salt of pevonedistat, a small-molecule inhibitor of the NEDD8-activating enzyme (NAE) used in preclinical research to probe ubiquitin-like protein modification, cell-cycle regulation, and anticancer mechanisms.

  • Potent, selective NAE inhibitor for mechanistic and pharmacology studies.
  • Hydrochloride salt with formula C21H26ClN5O4S.
  • Molecular weight 480.0 g/mol.
  • High purity suitable for research applications (~99%).
  • Soluble in DMSO; limited solubility in water.
  • Supplied as a 50 mg solid for small-scale experiments.

Catalog No. 50-002-42009 Supplier Medchemexpress LLC Supplier No. HY1048450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pevonedistat hydrochloride is the hydrochloride salt of pevonedistat, a small-molecule inhibitor of the NEDD8-activating enzyme (NAE) used in preclinical research to probe ubiquitin-like protein modification, cell-cycle regulation, and anticancer mechanisms.

  • Potent, selective NAE inhibitor for mechanistic and pharmacology studies.
  • Hydrochloride salt with formula C21H26ClN5O4S.
  • Molecular weight 480.0 g/mol.
  • High purity suitable for research applications (~99%).
  • Soluble in DMSO; limited solubility in water.
  • Supplied as a 50 mg solid for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.