Learn More
Selleck Chemical LLC Pevonedistat (MLN4924) 2mg 905579-51-3
SDP
Catalog No. 501947966
Encompass_Preferred
MLN4924 is a small molecule inhibitor of Nedd8 activating enzyme (NAE) with IC50 of 4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7966 Supplier Selleck Chemical LLC Supplier No. S71092MG
MLN4924 is a small molecule inhibitor of Nedd8 activating enzyme (NAE) with IC50 of 4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.