Learn More
Selleck Chemical LLC Pevonedistat (MLN4924) 5mg 905579-51-3
SDP
Supplier:  Selleck Chemical LLC S71095MG
Encompass_Preferred
MLN4924 is a small molecule inhibitor of Nedd8 activating enzyme (NAE) with IC50 of 4 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1533706