Learn More
MSC Pewag Chain Sling, Double-Leg, 5 ft, 7100 lb, 3/8 in
Catalog No. 5000531763
Encompass
This chain sling is engineered for demanding lifting applications, featuring a double-leg configuration with oblong and sling hooks. It measures 5 ft long and offers a robust vertical capacity of 7100 lb, maintaining this capacity at a 30-degree angle. Constructed from 3/8 in alloy steel, the chain is heat-treated and abrasion-resistant, ensuring durability and reliability in various industrial environments.
  • Double-leg design for balanced lifting
  • Oblong and sling hook attachments for secure loads
  • 5 ft length for flexible reach
  • 7100 lb vertical capacity for heavy loads
  • 3/8 in alloy steel chain
  • Grade 80 chain for heavy-duty performance
  • Heat-treated finish enhances strength
  • Abrasion-resistant for extended service life
  • Complies with OSHA, NACM, ISO 9001-2015, and ASTM A391/A391M standards
Catalog No. 50-005-31763 Supplier MSC Supplier No. 57292823
This chain sling is engineered for demanding lifting applications, featuring a double-leg configuration with oblong and sling hooks. It measures 5 ft long and offers a robust vertical capacity of 7100 lb, maintaining this capacity at a 30-degree angle. Constructed from 3/8 in alloy steel, the chain is heat-treated and abrasion-resistant, ensuring durability and reliability in various industrial environments.
  • Double-leg design for balanced lifting
  • Oblong and sling hook attachments for secure loads
  • 5 ft length for flexible reach
  • 7100 lb vertical capacity for heavy loads
  • 3/8 in alloy steel chain
  • Grade 80 chain for heavy-duty performance
  • Heat-treated finish enhances strength
  • Abrasion-resistant for extended service life
  • Complies with OSHA, NACM, ISO 9001-2015, and ASTM A391/A391M standards