Learn More
MSC Pewag Chain Sling, Grab Hook Single-Leg, 5 ft, 5200 lb, 9/32 in Grade 120
Catalog No. 5000530850
Encompass
The Pewag Chain Sling is designed with a Grab Hook and Single-Leg configuration. It features a 9/32 in, Grade 120 alloy steel chain, offering a vertical capacity of 5200 lb. This rust-resistant sling is 5 ft long and finished with a powder coating in light blue and gray, meeting EN 818 standards.
  • Grab hook and single-leg design
  • 5 ft length
  • 5200 lb vertical capacity
  • 9/32 in chain size
  • Grade 120 alloy steel chain
  • Rust-resistant
  • Powder coated finish
  • Meets EN 818 standards
  • Not adjustable
Catalog No. 50-005-30850 Supplier MSC Supplier No. 74939158
The Pewag Chain Sling is designed with a Grab Hook and Single-Leg configuration. It features a 9/32 in, Grade 120 alloy steel chain, offering a vertical capacity of 5200 lb. This rust-resistant sling is 5 ft long and finished with a powder coating in light blue and gray, meeting EN 818 standards.
  • Grab hook and single-leg design
  • 5 ft length
  • 5200 lb vertical capacity
  • 9/32 in chain size
  • Grade 120 alloy steel chain
  • Rust-resistant
  • Powder coated finish
  • Meets EN 818 standards
  • Not adjustable