Learn More
MSC Pewag Chain Sling, Single-Leg, Grab/Sling Hook, 10 ft, 7100 lb Capacity
Catalog No. 5000527866
Encompass
This single-leg chain sling from Pewag is engineered for demanding lifting applications. It features both a grab hook and a sling hook, providing versatility for various rigging setups. With a 10-foot length and a robust 7100 lb vertical capacity, this alloy steel chain sling is heat-treated for enhanced durability and abrasion resistance. It complies with multiple safety and quality standards, making it suitable for professional use.
  • Single-leg configuration.
  • Equipped with grab hook and sling hook.
  • 10 ft length.
  • 7100 lb vertical capacity.
  • Alloy steel construction.
  • Grade 80 chain sling.
  • Heat-treated finish.
  • Abrasion-resistant.
  • Complies with OSHA, NACM, ISO 9001-2015, and ASTM A391/A391M standards.
Catalog No. 50-005-27866 Supplier MSC Supplier No. 57292930
This single-leg chain sling from Pewag is engineered for demanding lifting applications. It features both a grab hook and a sling hook, providing versatility for various rigging setups. With a 10-foot length and a robust 7100 lb vertical capacity, this alloy steel chain sling is heat-treated for enhanced durability and abrasion resistance. It complies with multiple safety and quality standards, making it suitable for professional use.
  • Single-leg configuration.
  • Equipped with grab hook and sling hook.
  • 10 ft length.
  • 7100 lb vertical capacity.
  • Alloy steel construction.
  • Grade 80 chain sling.
  • Heat-treated finish.
  • Abrasion-resistant.
  • Complies with OSHA, NACM, ISO 9001-2015, and ASTM A391/A391M standards.