Learn More
Aviva Systems Biology Corp PEX10 Antibody - C-terminal region (ARP42997_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42997_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PEX10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PEX10. Alternate names: MGC1998, NALD, RNF69ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-58387