Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci PEX10 (CT) Peptide
SDP

Catalog No. 16595511
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-511 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-511 Supplier ProSci Supplier No. 9671P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

PEX10 (CT) Peptideis used for blocking the activity of thePEX10 (CT) Antibody

PEX10 is implicated in the biogenesis of peroxisomes. Mutations in this gene result in phenotypes within the Zellweger spectrum of peroxisomal biogenesis disorders, ranging from neonatal adrenoleukodystrophy to Zellweger syndrome.

Specifications

Gene ID (Entrez) 5192
Species Human
Accession Number O60683
Concentration 200 μg/mL
Content And Storage Store PEX10 (CT) Peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol PEX10
Quantity 0.05 mg
Sequence 10 amino acids near the carboxy terminus of PE x 10 (CT).
Synonym PEX10 Antibody: Peroxin-10, Peroxisomal biogenesis factor 10, Peroxisome assembly protein 10, RING finger protein 69, RNF69
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.