Learn More
Aviva Systems Biology Corp PEX19 Antibody - C-terminal region (ARP61545_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61545_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PEX19. It was validated on Western Blot. . Alternate names: D1S2223E, FLJ55296, HK33, PMP1, PMPI, PXF, PXMP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-405