Learn More
Aviva Systems Biology Corp Pex2 Antibody - C-terminal region (ARP56944_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56944_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pex2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1700016J08Rik, PXR2, Pex2, TRIP8bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-239