Learn More
ABclonal Technology PEX2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A153081000UL
Encompass_Preferred
This gene encodes an integral peroxisomal membrane protein required for peroxisome biogenesis. The protein is thought to be involved in peroxisomal matrix protein import. Mutations in this gene result in one form of Zellweger syndrome and infantile Refsum disease. Alternative splicing results in multiple transcript variants encoding the same protein.
Catalog No. 50-001-08151