Learn More
Aviva Systems Biology Corp PEX26 Antibody - middle region (ARP57068_P050) | 100uL
SDP
Catalog No. 5001462071
Encompass_Preferred
This is a rabbit polyclonal antibody against PEX26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PEX26. Alternate names: FLJ20695, PEX26M1T, Pex26pM1TApplicationWB. Species reactivity: Cow, Horse, Human, Pig
Catalog No. 50-014-62071 Supplier Aviva Systems Biology Corp Supplier No. ARP57068_P050
This is a rabbit polyclonal antibody against PEX26. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PEX26. Alternate names: FLJ20695, PEX26M1T, Pex26pM1TApplicationWB. Species reactivity: Cow, Horse, Human, Pig