Learn More
Aviva Systems Biology Corp PEX3 Antibody - N-terminal region (ARP46090_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46090_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PEX3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PEX3. Alternate names: TRG18ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59220