Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci PEX3 (CT) Peptide
SDP

Catalog No. 16595225
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-595-225 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-595-225 Supplier ProSci Supplier No. 9665P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

PEX3 (CT) Peptideis used for blocking the activity of thePEX3 (CT) Antibody

PEX3 Antibody: PEX3 is involved in peroxisome biosynthesis and integrity. It assembles membrane vesicles before the matrix proteins are translocated. As a docking factor for PEX19, PEX3 is necessary for the import of peroxisomal membrane proteins in the peroxisomes. Thus within the cell, PEX3 is stabilized by PEX19 preventing PEX3 aggregation. Mutations in the PEX19-binding domain of PEX3 reduce the affinity for PEX19 and destabilize PEX3.

Specifications

Gene ID (Entrez) 8504
Species Human
Accession Number P56589
Concentration 200 μg/mL
Content And Storage Store PEX3 (CT) Peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol PEX3
Quantity 0.05 mg
Sequence 12 amino acids near the carboxy terminus of PE x 3 (CT).
Synonym PEX3 Antibody: Peroxisomal biogenesis factor 3, Peroxin-3, Peroxisomal assembly protein PEX3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.