Learn More
Aviva Systems Biology Corp PEX7 Antibody - N-terminal region (ARP56091_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56091_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PEX7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PEX7. Alternate names: PTS2R, RCDP1, RDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61714