Learn More
Medchemexpress LLC HY-16749 100mg Medchemexpress, Pexidartinib CAS:1029044-16-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16749100MG
Encompass_Preferred
Medchemexpress, HY-16749 100mg Pexidartinib CAS:1029044-16-3 Pexidartinib (PLX-3397) is a potent, orally active, selective, and ATP-competitive colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit inhibitor, with IC50s of 20 and 10 nM, respectively. Pexidartinib (PLX-3397) exhibits 10- to 100-fold selectivity for c-Kit and CSF1R over other related kinases. Pexidartinib (PLX-3397) induces cell apoptosis and has anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9216