Learn More
Medchemexpress LLC HY-16749 200mg , Pexidartinib PLX-3397 CAS:1029044-16-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY16749200MG
Encompass_Preferred
Medchemexpress, HY-16749 200mg Pexidartinib PLX-3397 CAS:1029044-16-3 Formula:C20H15ClF3N5 IC50: 28 nM (c-Fms), 16 nM (c-Kit) Purity:98% Pexidartinib is a receptor tyrosine kinase (RTK) inhibitor of KIT , CSF1R and FLT3, with potential antineoplastic activity, with IC 50 of 28 and 16 nM for Fms and Kit, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2525