Learn More
Selleck Chemical LLC Pexidartinib Hydrochloride E6007-100MG
SDP
Catalog No. 5000041706
Encompass_Preferred
Pexidartinib Hydrochloride is an oral tyrosine kinase inhibitor of colony-stimulating factor 1 receptor (CSF1R) It significantly reduces macrophage levels in adipose tissue without altering overall myeloid cell counts in mice It also induces developmental and immunotoxic effects in zebrafish embryos by hyperactivating Wnt signaling
Catalog No. 50-000-41706 Supplier Selleck Chemical LLC Supplier No. E6007100MG
Pexidartinib Hydrochloride is an oral tyrosine kinase inhibitor of colony-stimulating factor 1 receptor (CSF1R) It significantly reduces macrophage levels in adipose tissue without altering overall myeloid cell counts in mice It also induces developmental and immunotoxic effects in zebrafish embryos by hyperactivating Wnt signaling