Learn More
Medchemexpress LLC Pexidartinib hydrochloride | 2040295-03-0 | 99.77% | 454.28 | 1 ML
SDP
Catalog No. 5000390722
Encompass_Preferred
Pexidartinib hydrochloride is a potent, orally active, selective, and ATP-competitive inhibitor of colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit. It exhibits high selectivity for c-Kit and CSF1R over other related kinases, induces cell apoptosis, and demonstrates anti-cancer activity.
  • Potent, orally active, selective, and ATP-competitive inhibitor
  • Induces cell apoptosis
  • Demonstrates anti-cancer activity
  • High selectivity for c-Kit and CSF1R over other related kinases
  • Can be used to deplete microglia cells in mice
  • Reduces tissue macrophage levels
Catalog No. 50-003-90722 Supplier Medchemexpress LLC Supplier No. HY16749A10MM/1ML
Pexidartinib hydrochloride is a potent, orally active, selective, and ATP-competitive inhibitor of colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit. It exhibits high selectivity for c-Kit and CSF1R over other related kinases, induces cell apoptosis, and demonstrates anti-cancer activity.
  • Potent, orally active, selective, and ATP-competitive inhibitor
  • Induces cell apoptosis
  • Demonstrates anti-cancer activity
  • High selectivity for c-Kit and CSF1R over other related kinases
  • Can be used to deplete microglia cells in mice
  • Reduces tissue macrophage levels