Learn More
Medchemexpress LLC Pexidartinib (hydrochloride) | 2040295-03-0 | 99.8% | 454.28 | 20 G
SDP
Catalog No. 5000391147
Encompass_Preferred
Pexidartinib hydrochloride is a potent, orally active, selective, and ATP-competitive colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit inhibitor. It exhibits 10- to 100-fold selectivity for CSF1R and c-Kit over other related kinases and induces cell apoptosis with anti-cancer activity.
  • Potent, orally active, selective, and ATP-competitive CSF1R and c-Kit inhibitor.
  • Exhibits 10- to 100-fold selectivity for CSF1R and c-Kit over other related kinases.
  • Induces cell apoptosis and has anti-cancer activity.
  • Can be used to deplete microglia cells in mice.
  • Reduces tissue macrophage levels without affecting glucose homeostasis in mice.
Catalog No. 50-003-91147 Supplier Medchemexpress LLC Supplier No. HY16749A20G
Pexidartinib hydrochloride is a potent, orally active, selective, and ATP-competitive colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit inhibitor. It exhibits 10- to 100-fold selectivity for CSF1R and c-Kit over other related kinases and induces cell apoptosis with anti-cancer activity.
  • Potent, orally active, selective, and ATP-competitive CSF1R and c-Kit inhibitor.
  • Exhibits 10- to 100-fold selectivity for CSF1R and c-Kit over other related kinases.
  • Induces cell apoptosis and has anti-cancer activity.
  • Can be used to deplete microglia cells in mice.
  • Reduces tissue macrophage levels without affecting glucose homeostasis in mice.