Learn More
Medchemexpress LLC Pexidartinib hydrochloride | 2040295-03-0 | 99.8% | 454.28 | 200 MG
SDP
Catalog No. 5000390897
Encompass_Preferred
Pexidartinib hydrochloride is a potent, orally active, selective, and ATP-competitive inhibitor of colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit. This compound induces cell apoptosis and demonstrates anti-cancer activity.
  • Potent, orally active, selective, and ATP-competitive inhibitor.
  • Targets colony stimulating factor 1 receptor (CSF1R) and c-Kit.
  • Exhibits high selectivity over other related kinases.
  • Induces cell apoptosis and demonstrates anti-cancer activity.
  • IC50 of 10 nM for c-Kit and 20 nM for cFMS.
Catalog No. 50-003-90897 Supplier Medchemexpress LLC Supplier No. HY16749A200MG
Pexidartinib hydrochloride is a potent, orally active, selective, and ATP-competitive inhibitor of colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit. This compound induces cell apoptosis and demonstrates anti-cancer activity.
  • Potent, orally active, selective, and ATP-competitive inhibitor.
  • Targets colony stimulating factor 1 receptor (CSF1R) and c-Kit.
  • Exhibits high selectivity over other related kinases.
  • Induces cell apoptosis and demonstrates anti-cancer activity.
  • IC50 of 10 nM for c-Kit and 20 nM for cFMS.