Learn More
Selleck Chemical LLC Pexidartinib Hydrochloride E6007-1G
SDP
Catalog No. 5000042341
Encompass_Preferred
Pexidartinib Hydrochloride is an oral tyrosine kinase inhibitor of colony-stimulating factor 1 receptor (CSF1R) It significantly reduces macrophage levels in adipose tissue without altering overall myeloid cell counts in mice It also induces developmental and immunotoxic effects in zebrafish embryos by hyperactivating Wnt signaling
Catalog No. 50-000-42341 Supplier Selleck Chemical LLC Supplier No. E60071G
Pexidartinib Hydrochloride is an oral tyrosine kinase inhibitor of colony-stimulating factor 1 receptor (CSF1R) It significantly reduces macrophage levels in adipose tissue without altering overall myeloid cell counts in mice It also induces developmental and immunotoxic effects in zebrafish embryos by hyperactivating Wnt signaling