Learn More
Selleck Chemical LLC Pexidartinib (PLX3397) 10mg 1029044-16-3
SDP
Supplier:  Selleck Chemical LLC S781810MG
Encompass_Preferred
Pexidartinib (PLX3397) is an oral, potent mutil-targeted receptor tyrosine kinase inhibitor of CSF-1R, Kit (c-Kit), and FLT3 with IC50 of 20 nM, 10 nM and 160 nM, respectively. Pexidartinib (PLX3397) induces apoptosis and necrosis with antitumor activity. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1124070