Learn More
Apexbio Technology LLC Pexidartinib (PLX3397) 1029044-16-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062764
Encompass_Preferred
Pexidartinib (PLX3397 CAS 1029044-16-3) is a small molecule inhibitor targeting colony-stimulating factor 1 receptor (CSF-1R) a receptor tyrosine kinase involved in macrophage recruitment and activation Through inhibition of CSF-1R activity this compound reduces macrophage infiltration and accumulation demonstrated by diminished macrophage levels in B16F10 melanoma models without affecting Gr-1 myeloid-derived suppressor cells In vivo studies using GL261 glioma models in C57 mice indicate decreased microglial infiltration upon oral dosing with PLX3397 Clinically combined treatment with Pexidartinib and paclitaxel is under evaluation in advanced solid tumors and breast cancer trials
Catalog No. 50-000-62764 Supplier Apexbio Technology LLC Supplier No. B58545.1
Pexidartinib (PLX3397 CAS 1029044-16-3) is a small molecule inhibitor targeting colony-stimulating factor 1 receptor (CSF-1R) a receptor tyrosine kinase involved in macrophage recruitment and activation Through inhibition of CSF-1R activity this compound reduces macrophage infiltration and accumulation demonstrated by diminished macrophage levels in B16F10 melanoma models without affecting Gr-1 myeloid-derived suppressor cells In vivo studies using GL261 glioma models in C57 mice indicate decreased microglial infiltration upon oral dosing with PLX3397 Clinically combined treatment with Pexidartinib and paclitaxel is under evaluation in advanced solid tumors and breast cancer trials