Learn More
Cayman Chemical PexIgnacetate 10mg
SDP
Catalog No. 502915220
Encompass_Preferred
An antimicrobial peptide; active against various clinical isolates of bacteria from patients with diabetic foot infections, including S. aureus, MRSA, and P. aeruginosa (MICs = 16-32, 16-32, and 8-16 UG/ml, respectively); decreases lethality in a mouse model of sepsis induced by A. baumannii at 1 mg/kg
Catalog No. 50-291-5220 Supplier Cayman Chemical Supplier No. 3692410 MG
An antimicrobial peptide; active against various clinical isolates of bacteria from patients with diabetic foot infections, including S. aureus, MRSA, and P. aeruginosa (MICs = 16-32, 16-32, and 8-16 UG/ml, respectively); decreases lethality in a mouse model of sepsis induced by A. baumannii at 1 mg/kg