Learn More
Medchemexpress LLC HY-16782 5mg Medchemexpress, Pexmetinib CAS:945614-12-0 Purity:>98%
SDP
Catalog No. 502081912
Encompass_Preferred
Medchemexpress, HY-16782 5mg Pexmetinib CAS:945614-12-0 Pexmetinib is a potent Tie-2 and p38 MAPK dual inhibitor, with IC50s of 1 nM, 35 nM and 26 nM for Tie-2, p38α and p38β, respectively, and can be used in the research of acute myeloid leukemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1912 Supplier Medchemexpress LLC Supplier No. HY167825MG
Medchemexpress, HY-16782 5mg Pexmetinib CAS:945614-12-0 Pexmetinib is a potent Tie-2 and p38 MAPK dual inhibitor, with IC50s of 1 nM, 35 nM and 26 nM for Tie-2, p38α and p38β, respectively, and can be used in the research of acute myeloid leukemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.