Learn More
Medchemexpress LLC Pexopiprant | 932708-14-0 | 99.4% | 456.27 | 100 MG
SDP
Catalog No. 5000344894
Encompass_Preferred
Pexopiprant is an oral antagonist of the prostaglandin D2 receptor 2 (DP2) with a Ki of less than 100 nM. It can be utilized in studies related to asthma.
  • Oral antagonist of prostaglandin D2 receptor 2 (DP2)
  • Ki of less than 100 nM
  • Suitable for asthma studies
Catalog No. 50-003-44894 Supplier Medchemexpress LLC Supplier No. HY10918610MG
Pexopiprant is an oral antagonist of the prostaglandin D2 receptor 2 (DP2) with a Ki of less than 100 nM. It can be utilized in studies related to asthma.
  • Oral antagonist of prostaglandin D2 receptor 2 (DP2)
  • Ki of less than 100 nM
  • Suitable for asthma studies